Assay Details
| Gene Symbol: Sema6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272024.1 | NP_001258953.1 | 6 - 7 | 789 | 105 | |
| NM_011351.2 | NP_035481.1 | 3 - 4 | 357 | 105 | ||
| NR_073549.1 | 4 - 5 | 636 | 105 | |||
| NR_073550.1 | 4 - 5 | 650 | 105 | |||
| NR_073551.1 | 5 - 6 | 699 | 105 | |||
| GenBank mRNA | AB013729.1 | - | 3 - 4 | 322 | 105 | |
| AF363972.1 | - | 3 - 4 | 390 | 105 | ||
| AF363973.1 | - | 3 - 4 | 390 | 105 | ||
| AK031131.1 | - | 4 - 5 | 548 | 105 | ||
| AK165085.1 | - | 4 - 5 | 563 | 105 | ||
| BC051979.1 | - | 5 - 6 | 605 | 105 | ||
| BC059007.1 | - | 6 - 7 | 704 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema6c
Genomic Map