Assay Details
| Gene Symbol: Frzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011356.4 | NP_035486.1 | 5 - 6 | 1417 | 61 | |
| GenBank mRNA | AK019093.2 | - | 5 - 6 | 1088 | 61 | |
| AK029941.1 | - | 5 - 6 | 1419 | 61 | ||
| BC016884.1 | - | 5 - 6 | 1264 | 4237554 | 61 | |
| U68058.1 | - | 5 - 6 | 873 | 61 | ||
| U88568.1 | - | 5 - 6 | 1225 | 61 | ||
| U91905.1 | - | 5 - 6 | 1659 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frzb
Genomic Map