| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100035670.mm.1 |
NM_011403.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035670.mm.1 |
NM_011403.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035670.mm.1 |
TC1100003833.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035669.mm.1 |
NM_011403.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035669.mm.1 |
NM_011403.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035669.mm.1 |
TC1100003833.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035668.mm.1 |
NM_011403.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035668.mm.1 |
NM_011403.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035668.mm.1 |
TC1100003833.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035667.mm.1 |
NM_011403.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035667.mm.1 |
NM_011403.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035667.mm.1 |
TC1100003833.mm.1 |
MTA TC |