Assay Details
| Gene Symbol: Slpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U73004.1 | - | 223 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the U73004.1 transcripts.
Set Membership: |
|

Slpi
Genomic Map