Assay Details
| Gene Symbol: Snai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011427.2 | NP_035557.1 | 1 - 2 | 145 | 79 | |
| GenBank mRNA | BC034857.1 | - | 1 - 2 | 145 | 5121591 | 79 |
| CT010204.1 | - | 1 - 2 | 82 | 79 | ||
| CT010370.1 | - | 1 - 2 | 82 | 79 | ||
| M95604.1 | - | 1 - 2 | 96 | 79 | ||
| X67253.1 | - | 1 - 2 | 102 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snai1
Genomic Map