Assay Details
| Gene Symbol: Tal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287388.1 | NP_001274317.1 | 3 - 4 | 658 | 97 | |
| NM_011527.3 | NP_035657.1 | 3 - 4 | 641 | 97 | ||
| GenBank mRNA | AK144164.1 | - | 2 - 3 | 612 | 97 | |
| AK157852.1 | - | 3 - 4 | 659 | 97 | ||
| BC063060.1 | - | 3 - 4 | 641 | 6826611 | 97 | |
| M59764.1 | - | 2 - 3 | 589 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tal1
Genomic Map