Assay Details
| Gene Symbol: Tnfsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011613.3 | NP_035743.2 | 2 - 3 | 534 | 66 | |
| GenBank mRNA | AB008426.1 | - | 2 - 3 | 380 | 66 | |
| AB032771.1 | - | 3 - 4 | 293 | 66 | ||
| AB032772.1 | - | 2 - 3 | 183 | 66 | ||
| AB036798.1 | - | 2 - 3 | 380 | 66 | ||
| AF013170.1 | - | 2 - 3 | 521 | 66 | ||
| AF019048.1 | - | 2 - 3 | 516 | 66 | ||
| AF053713.1 | - | 2 - 3 | 549 | 66 | ||
| AK159498.1 | - | 2 - 3 | 535 | 66 | ||
| AK159997.1 | - | 2 - 3 | 535 | 66 | ||
| BC125603.1 | - | 2 - 3 | 437 | 40130118 | 66 | |
| BC131970.1 | - | 2 - 3 | 437 | 40130247 | 66 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB032772.1 transcripts.
Set Membership: |
|

Tnfsf11
Genomic Map