Assay Details
| Gene Symbol: Trpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011643.2 | NP_035773.1 | 3 - 4 | 943 | 125 | |
| GenBank mRNA | BC137581.1 | - | 3 - 4 | 929 | 8860600 | 125 |
| U73625.1 | - | 3 - 4 | 943 | 125 | ||
| U95168.1 | - | 2 - 3 | 788 | 125 | ||
More Information
Set Membership: |
|

Trpc1
Genomic Map