Assay Details
| Gene Symbol: Trpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011643.2 | NP_035773.1 | 8 - 9 | 1961 | 95 | |
| GenBank mRNA | AF191551.1 | - | 7 - 8 | 1407 | 95 | |
| BC137581.1 | - | 8 - 9 | 1947 | 8860600 | 95 | |
| BC144726.1 | - | 7 - 8 | 1845 | 9053274 | 95 | |
| U73625.1 | - | 8 - 9 | 1961 | 95 | ||
| U95167.1 | - | 7 - 8 | 1859 | 95 | ||
| U95168.1 | - | 7 - 8 | 1806 | 95 | ||
| U95169.1 | - | 6 - 7 | 1704 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpc1
Genomic Map