Assay Details
| Gene Symbol: Trpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011643.2 | NP_035773.1 | 10 - 11 | 2277 | 67 | |
| GenBank mRNA | AF191551.1 | - | 9 - 10 | 1723 | 67 | |
| BC137581.1 | - | 10 - 11 | 2263 | 8860600 | 67 | |
| BC144726.1 | - | 9 - 10 | 2161 | 9053274 | 67 | |
| U40980.1 | - | 1 - 2 | 156 | 67 | ||
| U73625.1 | - | 10 - 11 | 2277 | 67 | ||
| U95167.1 | - | 9 - 10 | 2175 | 67 | ||
| U95168.1 | - | 9 - 10 | 2122 | 67 | ||
| U95169.1 | - | 8 - 9 | 2020 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpc1
Genomic Map