Assay Details
| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011881.3 | NP_036011.3 | 2 - 3 | 865 | 100 | |
| GenBank mRNA | BC096611.1 | - | 2 - 3 | 865 | 5357325 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|

Grk1
Genomic Map