Assay Details
| Gene Symbol: Mapk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168508.1 | NP_001161980.1 | 1 - 2 | 514 | 56 | |
| NM_011951.3 | NP_036081.1 | 1 - 2 | 514 | 56 | ||
| GenBank mRNA | AF128892.1 | - | 1 - 2 | 412 | 56 | |
| AK089059.1 | - | 1 - 2 | 428 | 56 | ||
| AK133684.1 | - | 1 - 2 | 427 | 56 | ||
| BC012235.1 | - | 1 - 2 | 514 | 4195842 | 56 | |
| BC085303.1 | - | 1 - 2 | 490 | 5389457 | 56 | |
| D83073.1 | - | 1 - 2 | 345 | 56 | ||
| U10871.1 | - | 1 - 2 | 412 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Inhibition of p38 mitogen-activated protein kinase enhances c-Jun N-terminal kinase activity: implication in inducible nitric oxide synthase expression.
- Authors
- Lahti A, Sareila O, Kankaanranta H, Moilanen E
- Journal
- Bmc Pharmacol - Page No 5 (Sun Jan 01 00:00:00 UTC 2006)
- Volume (Year)
- 6 (2006)
- Product Usage
- -

Mapk14
Genomic Map