Assay Details
| Gene Symbol: Mapk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168508.1 | NP_001161980.1 | 2 - 3 | 649 | 61 | |
| NM_001168513.1 | NP_001161985.1 | 2 - 3 | 250 | 61 | ||
| NM_001168514.1 | NP_001161986.1 | 3 - 4 | 694 | 61 | ||
| NM_011951.3 | NP_036081.1 | 2 - 3 | 649 | 61 | ||
| GenBank mRNA | AF128892.1 | - | 2 - 3 | 547 | 61 | |
| AK133684.1 | - | 2 - 3 | 562 | 61 | ||
| AK151348.1 | - | 2 - 3 | 251 | 61 | ||
| AK153025.1 | - | 2 - 3 | 251 | 61 | ||
| AK185139.1 | - | 2 - 3 | 166 | 61 | ||
| BC012235.1 | - | 2 - 3 | 649 | 4195842 | 61 | |
| BC018227.1 | - | 3 - 4 | 591 | 61 | ||
| BC085303.1 | - | 2 - 3 | 625 | 5389457 | 61 | |
| D83073.1 | - | 2 - 3 | 480 | 61 | ||
| U10871.1 | - | 2 - 3 | 547 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK151348.1,AK153025.1,NM_001168513.1,NM_001168514.1 transcripts.
Set Membership: |
|

Mapk14
Genomic Map