Assay Details
| Gene Symbol: Mapk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168513.1 | NP_001161985.1 | 9 - 10 | 761 | 65 | |
| NM_001168514.1 | NP_001161986.1 | 10 - 11 | 1205 | 65 | ||
| NM_011951.3 | NP_036081.1 | 9 - 10 | 1160 | 65 | ||
| GenBank mRNA | AK133684.1 | - | 9 - 10 | 1073 | 65 | |
| AK151348.1 | - | 9 - 10 | 762 | 65 | ||
| AK153025.1 | - | 9 - 10 | 762 | 65 | ||
| AK178357.1 | - | 1 - 2 | 102 | 65 | ||
| BC012235.1 | - | 9 - 10 | 1160 | 4195842 | 65 | |
| BC018227.1 | - | 10 - 11 | 1102 | 65 | ||
| U10871.1 | - | 9 - 10 | 1058 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk14
Genomic Map