Assay Details
| Gene Symbol: Mapk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168508.1 | NP_001161980.1 | 10 - 11 | 1244 | 92 | |
| NM_001168513.1 | NP_001161985.1 | 10 - 11 | 845 | 92 | ||
| NM_001168514.1 | NP_001161986.1 | 11 - 12 | 1289 | 92 | ||
| NM_011951.3 | NP_036081.1 | 10 - 11 | 1244 | 92 | ||
| GenBank mRNA | AF195850.1 | - | 7 - 8 | 567 | 92 | |
| AK089059.1 | - | 10 - 11 | 1158 | 92 | ||
| AK133684.1 | - | 10 - 11 | 1157 | 92 | ||
| AK151348.1 | - | 10 - 11 | 846 | 92 | ||
| AK153025.1 | - | 10 - 11 | 846 | 92 | ||
| BC012235.1 | - | 10 - 11 | 1244 | 4195842 | 92 | |
| BC018227.1 | - | 11 - 12 | 1186 | 92 | ||
| BC085303.1 | - | 10 - 11 | 1220 | 5389457 | 92 | |
| CX568915.1 | - | 9 - 10 | 638 | 92 | ||
| D83073.1 | - | 10 - 11 | 1075 | 92 | ||
| U10871.1 | - | 10 - 11 | 1142 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mapk14
Genomic Map