Assay Details
| Gene Symbol: Ppp2r5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081457.2 | NP_001074926.1 | 8 - 9 | 974 | 66 | |
| NM_001081458.2 | NP_001074927.1 | 8 - 9 | 974 | 66 | ||
| NM_001135001.1 | NP_001128473.1 | 10 - 11 | 1205 | 66 | ||
| NM_012023.3 | NP_036153.2 | 8 - 9 | 974 | 66 | ||
| GenBank mRNA | AB055635.1 | - | 8 - 9 | 824 | 66 | |
| AB055636.1 | - | 8 - 9 | 824 | 66 | ||
| AK031584.1 | - | 8 - 9 | 977 | 66 | ||
| AK129042.1 | - | 10 - 11 | 1077 | 66 | ||
| AK191098.1 | - | 3 - 4 | 190 | 66 | ||
| AK196453.1 | - | 3 - 4 | 190 | 66 | ||
| BC003979.1 | - | 8 - 9 | 934 | 3489999 | 66 | |
| U37353.1 | - | 8 - 9 | 803 | 66 | ||
More Information
Set Membership: |
|

Ppp2r5c
Genomic Map