Assay Details
| Gene Symbol: Atp1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013415.5 | NP_038201.1 | 2 - 3 | 828 | 91 | |
| GenBank mRNA | AK141786.1 | - | 2 - 3 | 829 | 91 | |
| AK158321.1 | - | 2 - 3 | 830 | 91 | ||
| BC034586.1 | - | 2 - 3 | 810 | 91 | ||
| BC042467.1 | - | 2 - 3 | 697 | 4500972 | 91 | |
| BC058763.1 | - | 2 - 3 | 786 | 6490870 | 91 | |
| X16645.1 | - | 2 - 3 | 384 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1b2
Genomic Map