Assay Details
| Gene Symbol: Atp1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013415.5 | NP_038201.1 | 3 - 4 | 931 | 71 | |
| GenBank mRNA | AK141786.1 | - | 3 - 4 | 932 | 71 | |
| AK158321.1 | - | 3 - 4 | 934 | 71 | ||
| BC034586.1 | - | 3 - 4 | 913 | 71 | ||
| BC042467.1 | - | 3 - 4 | 800 | 4500972 | 71 | |
| BC058763.1 | - | 3 - 4 | 889 | 6490870 | 71 | |
| X16645.1 | - | 3 - 4 | 487 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1b2
Genomic Map