Assay Details
| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013566.2 | NP_038594.2 | 1 - 2 | 86 | 80 | |
| GenBank mRNA | AK078871.1 | - | 1 - 2 | 87 | 80 | |
| AK144773.1 | - | 1 - 2 | 58 | 80 | ||
| AK154766.1 | - | 1 - 2 | 87 | 80 | ||
| AK155207.1 | - | 1 - 2 | 53 | 80 | ||
| AK155594.1 | - | 1 - 2 | 58 | 80 | ||
| BC011184.1 | - | 1 - 2 | 59 | 4187781 | 80 | |
| M68903.1 | - | 1 - 2 | 37 | 80 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK078871.1,AK154766.1,AK155207.1,BC011184.1,M68903.1,NM_013566.2 transcripts.
Set Membership: |
|

Itgb7
Genomic Map