Assay Details
| Gene Symbol: Slc11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013612.2 | NP_038640.2 | 2 - 3 | 256 | 68 | |
| GenBank mRNA | AK090303.1 | - | 2 - 3 | 257 | 68 | |
| AK144237.1 | - | 2 - 3 | 239 | 68 | ||
| AK171343.1 | - | 2 - 3 | 185 | 68 | ||
| AK171393.1 | - | 2 - 3 | 237 | 68 | ||
| BC109137.1 | - | 2 - 3 | 170 | 40039390 | 68 | |
| BC109138.1 | - | 2 - 3 | 170 | 40039397 | 68 | |
| L13732.1 | - | 2 - 3 | 234 | 68 | ||
| X75355.1 | - | 2 - 3 | 215 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc11a1
Genomic Map