Assay Details
| Gene Symbol: Pklr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013631.2 | NP_038659.2 | 1 - 2 | 144 | 105 | |
| GenBank mRNA | BC152327.1 | - | 1 - 2 | 128 | 40141144 | 105 |
| BC152542.1 | - | 1 - 2 | 128 | 105 | ||
| BY107231.1 | - | 1 - 2 | 112 | 105 | ||
| D63764.1 | - | 1 - 2 | 144 | 105 | ||
| S79731.1 | - | 1 - 2 | 144 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pklr
Genomic Map