| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800006874.mm.1 |
NM_013641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006874.mm.1 |
NM_013641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006874.mm.1 |
TC0800000931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006876.mm.1 |
NM_013641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006876.mm.1 |
NM_013641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006876.mm.1 |
TC0800000931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006877.mm.1 |
NM_013641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006877.mm.1 |
NM_013641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006877.mm.1 |
TC0800000931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006875.mm.1 |
NM_013641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006875.mm.1 |
NM_013641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006875.mm.1 |
TC0800000931.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006878.mm.1 |
NM_013641.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006878.mm.1 |
NM_013641.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006878.mm.1 |
TC0800000931.mm.1 |
MTA TC |