Assay Details
| Gene Symbol: Rora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289916.1 | NP_001276845.1 | 4 - 5 | 828 | 84 | |
| NM_013646.2 | NP_038674.1 | 5 - 6 | 837 | 84 | ||
| GenBank mRNA | AK043990.1 | - | 1 - 2 | 197 | 84 | |
| AK149294.1 | - | 4 - 5 | 718 | 84 | ||
| AK155827.1 | - | 4 - 5 | 779 | 84 | ||
| BC003757.1 | - | 4 - 5 | 777 | 3592667 | 84 | |
| U53228.1 | - | 5 - 6 | 821 | 84 | ||
| Y08640.1 | - | 4 - 5 | 983 | 84 | ||
| Z82994.1 | - | 4 - 5 | 816 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rora
Genomic Map