Assay Details
| Gene Symbol: Ccl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013654.3 | NP_038682.1 | 1 - 2 | 148 | 122 | |
| GenBank mRNA | AF128193.1 | - | 1 - 2 | 113 | 122 | |
| AF128194.1 | - | 1 - 2 | 113 | 122 | ||
| AK078827.1 | - | 1 - 2 | 149 | 122 | ||
| BC061126.1 | - | 1 - 2 | 146 | 30276350 | 122 | |
| BY163435.1 | - | 1 - 2 | 150 | 122 | ||
| L04694.1 | - | 1 - 2 | 110 | 122 | ||
| S50588.1 | - | 1 - 2 | 96 | 122 | ||
| S71251.1 | - | 1 - 2 | 128 | 122 | ||
| Z12297.1 | - | 1 - 2 | 96 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl7
Genomic Map