Assay Details
| Gene Symbol: Sema4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163489.1 | NP_001156961.1 | 3 - 4 | 362 | 96 | |
| NM_001163490.1 | NP_001156962.1 | 3 - 4 | 441 | 96 | ||
| NM_001163491.1 | NP_001156963.1 | 3 - 4 | 400 | 96 | ||
| NM_013658.3 | NP_038686.3 | 3 - 4 | 540 | 96 | ||
| GenBank mRNA | AK035918.1 | - | 2 - 3 | 355 | 96 | |
| AK077976.1 | - | 3 - 4 | 520 | 96 | ||
| AK152728.1 | - | 3 - 4 | 532 | 96 | ||
| AK153164.1 | - | 3 - 4 | 532 | 96 | ||
| AK154392.1 | - | 3 - 4 | 501 | 96 | ||
| BB840212.1 | - | 3 - 4 | 401 | 96 | ||
| BC025800.1 | - | 3 - 4 | 458 | 5102015 | 96 | |
| X85991.1 | - | 3 - 4 | 362 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4a
Genomic Map