Assay Details
| Gene Symbol: Sema4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163489.1 | NP_001156961.1 | 13 - 14 | 1655 | 65 | |
| NM_001163490.1 | NP_001156962.1 | 13 - 14 | 1734 | 65 | ||
| NM_001163491.1 | NP_001156963.1 | 13 - 14 | 1693 | 65 | ||
| NM_013658.3 | NP_038686.3 | 13 - 14 | 1833 | 65 | ||
| GenBank mRNA | AK035918.1 | - | 12 - 13 | 1695 | 65 | |
| AK077976.1 | - | 13 - 14 | 1814 | 65 | ||
| AK154392.1 | - | 13 - 14 | 1794 | 65 | ||
| BC025800.1 | - | 13 - 14 | 1751 | 5102015 | 65 | |
| X85991.1 | - | 13 - 14 | 1655 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4a
Genomic Map