Assay Details
| Gene Symbol: Tap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161730.1 | NP_001155202.1 | 2 - 3 | 963 | 88 | |
| NM_013683.2 | NP_038711.2 | 2 - 3 | 963 | 88 | ||
| GenBank mRNA | AK040629.1 | - | 2 - 3 | 792 | 88 | |
| AK151481.1 | - | 2 - 3 | 718 | 88 | ||
| AK152953.1 | - | 2 - 3 | 663 | 88 | ||
| AK153890.1 | - | 2 - 3 | 967 | 88 | ||
| AK162731.1 | - | 2 - 3 | 763 | 88 | ||
| AK166046.1 | - | 2 - 3 | 964 | 88 | ||
| AK167485.1 | - | 2 - 3 | 965 | 88 | ||
| AK171363.1 | - | 2 - 3 | 718 | 88 | ||
| BC013802.1 | - | 2 - 3 | 374 | 3257566 | 88 | |
| BC024897.1 | - | 2 - 3 | 486 | 3257734 | 88 | |
| M55637.1 | - | 2 - 3 | 355 | 88 | ||
| U60018.1 | - | 2 - 3 | 639 | 88 | ||
| U60019.1 | - | 2 - 3 | 639 | 88 | ||
| U60020.1 | - | 2 - 3 | 639 | 88 | ||
| U60021.1 | - | 2 - 3 | 639 | 88 | ||
| U60022.1 | - | 2 - 3 | 639 | 88 | ||
| U60023.1 | - | 2 - 3 | 639 | 88 | ||
| X59615.1 | - | 2 - 3 | 198 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tap1
Genomic Map