Assay Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145959.1 | NP_001139431.1 | 4 - 5 | 473 | 73 | |
| NM_013864.2 | NP_038892.1 | 5 - 6 | 515 | 73 | ||
| GenBank mRNA | AB033921.1 | - | 5 - 6 | 378 | 73 | |
| AK076514.1 | - | 2 - 3 | 149 | 73 | ||
| AK154597.1 | - | 5 - 6 | 383 | 73 | ||
| AK158900.1 | - | 4 - 5 | 382 | 73 | ||
| AK168011.1 | - | 4 - 5 | 340 | 73 | ||
| AK173136.1 | - | 2 - 3 | 156 | 73 | ||
| BC012963.1 | - | 5 - 6 | 382 | 4211421 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndrg2
Genomic Map