Assay Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145959.1 | NP_001139431.1 | 11 - 12 | 941 | 95 | |
| NM_013864.2 | NP_038892.1 | 12 - 13 | 983 | 95 | ||
| GenBank mRNA | AB033921.1 | - | 12 - 13 | 846 | 95 | |
| AK076514.1 | - | 9 - 10 | 617 | 95 | ||
| AK154597.1 | - | 12 - 13 | 851 | 95 | ||
| AK158900.1 | - | 11 - 12 | 850 | 95 | ||
| AK168011.1 | - | 11 - 12 | 808 | 95 | ||
| AK173136.1 | - | 9 - 10 | 624 | 95 | ||
| BC012963.1 | - | 12 - 13 | 850 | 4211421 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndrg2
Genomic Map