Assay Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145959.1 | NP_001139431.1 | 12 - 13 | 991 | 70 | |
| NM_013864.2 | NP_038892.1 | 13 - 14 | 1033 | 70 | ||
| GenBank mRNA | AB033921.1 | - | 13 - 14 | 896 | 70 | |
| AK076514.1 | - | 10 - 11 | 667 | 70 | ||
| AK154597.1 | - | 13 - 14 | 901 | 70 | ||
| AK158900.1 | - | 12 - 13 | 900 | 70 | ||
| AK168011.1 | - | 12 - 13 | 858 | 70 | ||
| AK173136.1 | - | 10 - 11 | 674 | 70 | ||
| BC012963.1 | - | 13 - 14 | 900 | 4211421 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndrg2
Genomic Map