Assay Details
| Gene Symbol: Tcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130458.1 | NP_001123930.1 | 5 - 6 | 787 | 128 | |
| NM_001130459.1 | NP_001123931.1 | 5 - 6 | 756 | 128 | ||
| NM_015749.3 | NP_056564.1 | 5 - 6 | 790 | 128 | ||
| GenBank mRNA | AF090686.1 | - | 4 - 5 | 717 | 128 | |
| AK133707.1 | - | 5 - 6 | 752 | 128 | ||
| AK143761.1 | - | 5 - 6 | 758 | 128 | ||
| AK147176.1 | - | 5 - 6 | 761 | 128 | ||
| AK161618.1 | - | 5 - 6 | 757 | 128 | ||
| AK170401.1 | - | 5 - 6 | 776 | 128 | ||
| BC003720.1 | - | 4 - 5 | 663 | 3601531 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcn2
Genomic Map