Assay Details
| Gene Symbol: Npy5r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016708.3 | NP_057917.2 | 1 - 2 | 128 | 148 | |
| GenBank mRNA | AB001346.1 | - | 1 - 2 | 131 | 148 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB001346.1,NM_016708.3 transcripts.
Set Membership: |
|

Npy5r
Genomic Map