Assay Details
| Gene Symbol: Bnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008238.3 | NP_001008239.1 | 6 - 7 | 848 | 94 | |
| NM_016787.4 | NP_058067.2 | 6 - 7 | 848 | 94 | ||
| GenBank mRNA | AF035207.1 | - | 6 - 7 | 700 | 94 | |
| AF400107.1 | - | 5 - 6 | 588 | 94 | ||
| AK014659.1 | - | 6 - 7 | 800 | 94 | ||
| AK054288.1 | - | 6 - 7 | 814 | 94 | ||
| AK159745.1 | - | 6 - 7 | 811 | 94 | ||
| AK161116.1 | - | 6 - 7 | 801 | 94 | ||
| AK169609.1 | - | 6 - 7 | 833 | 94 | ||
| AY591757.1 | - | 5 - 6 | 567 | 94 | ||
| AY591758.1 | - | 5 - 6 | 567 | 94 | ||
| BC012670.1 | - | 6 - 7 | 807 | 4037627 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bnip2
Genomic Map