Assay Details
| Gene Symbol: Ccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016892.3 | NP_058588.1 | 4 - 5 | 513 | 127 | |
| GenBank mRNA | AF121906.1 | - | 4 - 5 | 426 | 127 | |
| AF173379.1 | - | 4 - 5 | 486 | 127 | ||
| AK010264.1 | - | 1 - 2 | 122 | 127 | ||
| AK050072.1 | - | 4 - 5 | 462 | 127 | ||
| BC026938.1 | - | 4 - 5 | 480 | 4024140 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccs
Genomic Map