Assay Details
| Gene Symbol: Apom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018816.1 | NP_061286.1 | 3 - 4 | 377 | 68 | |
| GenBank mRNA | AF207820.1 | - | 3 - 4 | 377 | 68 | |
| AK004530.1 | - | 3 - 4 | 422 | 68 | ||
| BC021597.1 | - | 3 - 4 | 414 | 5100655 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apom
Genomic Map