Assay Details
| Gene Symbol: Cd86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019388.3 | NP_062261.3 | 3 - 4 | 517 | 130 | |
| GenBank mRNA | AF065897.1 | - | 3 - 4 | 433 | 130 | |
| AF065898.1 | - | 3 - 4 | 433 | 130 | ||
| AF065899.1 | - | 3 - 4 | 433 | 130 | ||
| AF065900.1 | - | 3 - 4 | 433 | 130 | ||
| AK079513.1 | - | 3 - 4 | 518 | 130 | ||
| AK170054.1 | - | 3 - 4 | 518 | 130 | ||
| AK172548.1 | - | 3 - 4 | 520 | 130 | ||
| L25606.1 | - | 3 - 4 | 499 | 130 | ||
| S70108.1 | - | 3 - 4 | 482 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd86
Genomic Map