Assay Details
| Gene Symbol: Il17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019508.1 | NP_062381.1 | 1 - 2 | 75 | ||
| GenBank mRNA | AF184970.1 | - | 1 - 2 | 75 | ||
| AF218726.1 | - | 1 - 2 | 75 | |||
| AF250308.1 | - | 1 - 2 | 75 | |||
| BC002271.1 | - | 1 - 2 | 3496106 | 75 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF184970.1,AF218726.1,AF250308.1,BC002271.1,NM_019508.1 transcripts.
Set Membership: |
|

Il17b
Genomic Map