Assay Details
| Gene Symbol: Pfkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291071.1 | NP_001278000.1 | 14 - 15 | 1530 | 75 | |
| NM_019703.4 | NP_062677.1 | 14 - 15 | 1530 | 75 | ||
| GenBank mRNA | AF123533.1 | - | 14 - 15 | 1495 | 75 | |
| AK004790.1 | - | 14 - 15 | 1484 | 75 | ||
| AK049841.1 | - | 14 - 15 | 1484 | 75 | ||
| AK054193.1 | - | 14 - 15 | 1487 | 75 | ||
| AK076803.1 | - | 14 - 15 | 1485 | 75 | ||
| AK078254.1 | - | 14 - 15 | 1485 | 75 | ||
| AK098162.1 | - | 14 - 15 | 1484 | 75 | ||
| AK152670.1 | - | 14 - 15 | 1485 | 75 | ||
| AK154125.1 | - | 14 - 15 | 1485 | 75 | ||
| AK170624.1 | - | 14 - 15 | 1485 | 75 | ||
| AK171062.1 | - | 14 - 15 | 1485 | 75 | ||
| AY779275.1 | - | 14 - 15 | 1447 | 75 | ||
| AY779276.1 | - | 14 - 15 | 1447 | 75 | ||
| BC006926.1 | - | 14 - 15 | 1469 | 2655383 | 75 | |
| CT010268.1 | - | 14 - 15 | 1438 | 75 | ||
| Y19008.1 | - | 14 - 15 | 1487 | 75 | ||
More Information
Set Membership: |
|

Pfkp
Genomic Map