Assay Details
| Gene Symbol: Pfkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291071.1 | NP_001278000.1 | 19 - 20 | 2106 | 97 | |
| NM_019703.4 | NP_062677.1 | 19 - 20 | 2106 | 97 | ||
| GenBank mRNA | AF123533.1 | - | 19 - 20 | 2071 | 97 | |
| AK004790.1 | - | 19 - 20 | 2060 | 97 | ||
| AK047303.1 | - | 1 - 2 | 105 | 97 | ||
| AK049841.1 | - | 19 - 20 | 2060 | 97 | ||
| AK076803.1 | - | 19 - 20 | 2061 | 97 | ||
| AK078254.1 | - | 19 - 20 | 2061 | 97 | ||
| AK098162.1 | - | 19 - 20 | 2060 | 97 | ||
| AK152670.1 | - | 19 - 20 | 2061 | 97 | ||
| AK154125.1 | - | 19 - 20 | 2061 | 97 | ||
| AK170624.1 | - | 19 - 20 | 2061 | 97 | ||
| AK171062.1 | - | 19 - 20 | 2061 | 97 | ||
| AY779275.1 | - | 19 - 20 | 2023 | 97 | ||
| AY779276.1 | - | 19 - 20 | 2023 | 97 | ||
| BC006926.1 | - | 19 - 20 | 2045 | 2655383 | 97 | |
| CT010268.1 | - | 19 - 20 | 2014 | 97 | ||
| Y19008.1 | - | 19 - 20 | 2063 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkp
Genomic Map