Assay Details
| Gene Symbol: Tubd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199045.1 | NP_001185974.1 | 9 - 10 | 1744 | 71 | |
| NM_001199046.1 | NP_001185975.1 | 9 - 10 | 1638 | 71 | ||
| NM_001199047.1 | NP_001185976.1 | 8 - 9 | 1651 | 71 | ||
| NM_019756.3 | NP_062730.1 | 8 - 9 | 1545 | 71 | ||
| GenBank mRNA | AB024329.1 | - | 7 - 8 | 1319 | 71 | |
| AB031043.1 | - | 8 - 9 | 1350 | 71 | ||
| AF081568.1 | - | 8 - 9 | 1385 | 71 | ||
| AK030214.1 | - | 9 - 10 | 1744 | 71 | ||
| AK169454.1 | - | 8 - 9 | 1546 | 71 | ||
| AK171546.1 | - | 8 - 9 | 1512 | 71 | ||
| AK216514.1 | - | 1 - 2 | 49 | 71 | ||
| BC026465.1 | - | 8 - 9 | 1528 | 4484099 | 71 | |
| BC132325.1 | - | 8 - 9 | 1378 | 40130602 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27989558 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098578 | BC026465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098578 | AK170304.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098578 | AK171546.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098578 | AK216514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098578 | AK030214.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958662 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303484 | AK076507.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303484 | NM_007410.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461802 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13649 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107528 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098578 | BC026465.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098578 | AK170304.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098578 | AK171546.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098578 | AK216514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2098578 | AK030214.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302056 | BC026465.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302056 | BC132325.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302056 | AK169454.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302056 | AK170304.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302056 | AK171546.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27989567 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07428895 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07428930 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07428915 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02385379 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107528 | - | - |
| Whole Mouse Genome G4122A | A_51_P302056 | BC026465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302056 | BC132325.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302056 | AK169454.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302056 | AK170304.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302056 | AK171546.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136779 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07428915 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107528 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09665915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09190959 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09665920 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27989558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27989640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09414730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09531899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09190959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09266145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09730209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09334018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09003760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09003761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09293246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09553888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27989586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09665915 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379969 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379973 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379971 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379972 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379976 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379974 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379978 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421748_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379968 | - | - |
| Mouse Genome 430 2.0 Array | 1421748_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421748_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56427_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56427_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_001199045.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_001199045.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_001199046.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_001199046.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_001199047.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_001199047.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_019756.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | NM_019756.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012352.mm.1 | TC1100001317.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_001199045.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_001199045.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_001199046.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_001199046.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_001199047.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_001199047.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_019756.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | NM_019756.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100012351.mm.1 | TC1100001317.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421748_a_at | - | - |
| Mouse Expression Set 430 | 1421748_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421748_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421748_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421748_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421748_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5340093 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5340093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290059 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2464061 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2506229 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2464061 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2506229 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001490.1_34-S | - | - |
| mouse-6 v1.1 (Illumina) | scl41095.10.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290059 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340093 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41095.10.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001490.1_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340093 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290059 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2464061 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2506229 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2464061 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2506229 | - | - |

Tubd1
Genomic Map