Assay Details
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021340.4 | NP_067315.1 | 3 - 4 | 450 | 125 | |
| GenBank mRNA | AF076930.1 | - | 3 - 4 | 370 | 125 | |
| AK044450.1 | - | 3 - 4 | 402 | 125 | ||
| AK044649.1 | - | 3 - 4 | 401 | 125 | ||
| AK149272.1 | - | 3 - 4 | 401 | 125 | ||
| BC082611.1 | - | 3 - 4 | 387 | 125 | ||
| BC086689.1 | - | 3 - 4 | 377 | 125 | ||
| BC119055.1 | - | 3 - 4 | 384 | 8733804 | 125 | |
| CK622127.1 | - | 3 - 4 | 450 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgr
Genomic Map