Assay Details
| Gene Symbol: Efemp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164352.1 | NP_001157824.1 | 4 - 5 | 817 | 69 | |
| NM_021474.3 | NP_067449.3 | 5 - 6 | 576 | 69 | ||
| GenBank mRNA | AF104223.1 | - | 4 - 5 | 531 | 69 | |
| AF109122.1 | - | 4 - 5 | 790 | 69 | ||
| AK075585.1 | - | 5 - 6 | 565 | 69 | ||
| BC012269.1 | - | 4 - 5 | 714 | 3980048 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efemp2
Genomic Map