| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900000880.mm.1 |
NM_001164352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000880.mm.1 |
NM_001164352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000880.mm.1 |
NM_021474.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000880.mm.1 |
NM_021474.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000880.mm.1 |
TC1900000071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000879.mm.1 |
NM_001164352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000879.mm.1 |
NM_001164352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000879.mm.1 |
NM_021474.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000879.mm.1 |
NM_021474.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000879.mm.1 |
TC1900000071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900000878.mm.1 |
NM_001164352.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000878.mm.1 |
NM_001164352.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000878.mm.1 |
NM_021474.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000878.mm.1 |
NM_021474.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900000878.mm.1 |
TC1900000071.mm.1 |
MTA TC |