Assay Details
| Gene Symbol: Cxcl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012477.2 | NP_001012495.1 | 1 - 2 | 180 | 122 | |
| NM_013655.4 | NP_038683.1 | 1 - 2 | 180 | 122 | ||
| NM_021704.3 | NP_068350.1 | 1 - 2 | 180 | 122 | ||
| GenBank mRNA | AK045092.1 | - | 1 - 2 | 139 | 122 | |
| AK075596.1 | - | 1 - 2 | 139 | 122 | ||
| AK157553.1 | - | 1 - 2 | 141 | 122 | ||
| BB610070.1 | - | 1 - 2 | 136 | 122 | ||
| BB840704.1 | - | 1 - 2 | 139 | 122 | ||
| BC006640.1 | - | 1 - 2 | 128 | 3483088 | 122 | |
| BC020297.1 | - | 1 - 2 | 128 | 122 | ||
| BC029335.1 | - | 1 - 2 | 128 | 122 | ||
| BC046827.1 | - | 1 - 2 | 130 | 122 | ||
| D21072.1 | - | 1 - 2 | 117 | 122 | ||
| D43804.1 | - | 1 - 2 | 137 | 122 | ||
| D43805.1 | - | 1 - 2 | 137 | 122 | ||
| L12029.1 | - | 1 - 2 | 140 | 122 | ||
| L12030.1 | - | 1 - 2 | 135 | 122 | ||
| S74318.1 | - | 1 - 2 | 125 | 122 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK045092.1,AK075596.1,AK157553.1,BC006640.1,D21072.1,D43804.1,D43805.1,L12029.1,L12030.1,NM_001012477.2,NM_013655.4,NM_021704.3,S74318.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Pro-inflammatory properties of stromal cell-derived factor-1 (CXCL12) in collagen-induced arthritis.
- Authors
- De Klerck B, Geboes L, Hatse S, Kelchtermans H, Meyvis Y, Vermeire K, Bridger G, Billiau A, Schols D, Matthys P
- Journal
- Arthritis Res Ther - Page No R1208 (Sat Jan 01 00:00:00 UTC 2005)
- Volume (Year)
- 7 (2005)
- Product Usage
- -

Cxcl12
Genomic Map