Assay Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023232.3 | NP_075721.3 | 4 - 5 | 461 | 87 | |
| GenBank mRNA | AF203914.1 | - | 4 - 5 | 427 | 87 | |
| AK002887.1 | - | 4 - 5 | 431 | 87 | ||
| AK012760.1 | - | 4 - 5 | 459 | 87 | ||
| AK077198.1 | - | 4 - 5 | 442 | 87 | ||
| AK133194.1 | - | 441 | 87 | |||
| BC054109.1 | - | 4 - 5 | 455 | 30013833 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Diablo
Genomic Map