Assay Details
| Gene Symbol: Ndufa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023312.2 | NP_075801.1 | 4 - 5 | 1018 | 75 | |
| GenBank mRNA | AF286698.1 | - | 3 - 4 | 253 | 75 | |
| AK002999.1 | - | 3 - 4 | 257 | 75 | ||
| AK003012.1 | - | 3 - 4 | 271 | 75 | ||
| AK010517.1 | - | 3 - 4 | 257 | 75 | ||
| AK012422.1 | - | 3 - 4 | 257 | 75 | ||
| BC034899.1 | - | 4 - 5 | 1018 | 1447015 | 75 | |
| BC037149.1 | - | 3 - 4 | 261 | 5149812 | 75 | |
| BC055435.1 | - | 3 - 4 | 267 | 4219489 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cell growth
cellular response to interferon-beta
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein import into mitochondrial inner membrane
positive regulation of protein catabolic process
negative regulation of transcription, DNA-templated
oxidation-reduction process
cellular response to retinoic acid
reactive oxygen species metabolic process
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
NADH dehydrogenase activity
ATP binding
NADH dehydrogenase (ubiquinone) activity
negative regulation of cell growth
cellular response to interferon-beta
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein import into mitochondrial inner membrane
positive regulation of protein catabolic process
negative regulation of transcription, DNA-templated
oxidation-reduction process
cellular response to retinoic acid
reactive oxygen species metabolic process
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
NADH dehydrogenase activity
ATP binding
NADH dehydrogenase (ubiquinone) activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948623 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948622 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948621 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948616 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894674 | NM_146448.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49766 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113452 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25948616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25948620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25948619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25948624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25948622 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06547004 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113452 | - | - |
| Whole Mouse Genome G4122A | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | AK003012.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | AK010517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | AK012422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | BC034899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381360 | BC037149.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116295 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140147 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113452 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07351539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07351531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07351554 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948621 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948624 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948622 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948616 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25948623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25948541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25948615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25948622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25948616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25948624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25948619 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10579122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579124 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430713_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572251 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10579119 | - | - |
| Mouse Genome 430 2.0 Array | 1430713_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1430713_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67184_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67184_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800017896.mm.1 | NM_023312.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800017896.mm.1 | NM_023312.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800017896.mm.1 | TC0800002376.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430713_s_at | - | - |
| Mouse Expression Set 430 | 1430713_PM_s_at | - | - |
| Murine Genome U74v2 Set | 93764_at | - | - |
| Murine Genome U74v2 Set | 93764_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430713_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430713_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430713_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430713_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101500132 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6220021 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101500132 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006220021 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649937 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2520382 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649937 | - | - |
| mouse-6 v1.1 (Illumina) | scl34723.1_7-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0067184.2_306-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101500132 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6220021 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067184.2_306-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101500132 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220021 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2649937 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2649937 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0067184.2_306-S | - | - |

Ndufa13
Genomic Map