Assay Details
| Gene Symbol: Npy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023456.2 | NP_075945.1 | 2 - 3 | 259 | 65 | |
| GenBank mRNA | AF273768.1 | - | 1 - 2 | 186 | 65 | |
| AF286198.1 | - | 1 - 2 | 186 | 65 | ||
| AK002982.1 | - | 2 - 3 | 272 | 65 | ||
| BC043012.1 | - | 2 - 3 | 259 | 5683102 | 65 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Citations (2)
Fibroblast growth factor 21 corrects obesity in mice.
- Authors
- Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
- Journal
- Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 UTC 2008)
- Volume (Year)
- 149 (2008)
- Product Usage
- -
Hyperphagia-mediated obesity in transgenic mice misexpressing the RNA-editing enzyme ADAR2.
- Authors
- Singh M, Kesterson RA, Jacobs MM, Joers JM, Gore JC, Emeson RB
- Journal
- J Biol Chem - Page No 22448 (Fri Aug 03 00:00:00 UTC 2007)
- Volume (Year)
- 282 (2007)
- Product Usage
- -

Npy
Genomic Map