Assay Details
| Gene Symbol: Atp6v1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023721.2 | NP_076210.1 | 2 - 3 | 339 | 77 | |
| GenBank mRNA | AF298810.1 | - | 2 - 3 | 160 | 77 | |
| AK146448.1 | - | 2 - 3 | 242 | 77 | ||
| AK147910.1 | - | 256 | 77 | |||
| AK150981.1 | - | 2 - 3 | 228 | 77 | ||
| AK152367.1 | - | 2 - 3 | 228 | 77 | ||
| AK153250.1 | - | 2 - 3 | 228 | 77 | ||
| AK165761.1 | - | 2 - 3 | 233 | 77 | ||
| BC033457.1 | - | 2 - 3 | 201 | 3991107 | 77 | |
| BY096594.1 | - | 2 - 3 | 341 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1d
Genomic Map