Assay Details
| Gene Symbol: Atp6v1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023721.2 | NP_076210.1 | 5 - 6 | 533 | 65 | |
| GenBank mRNA | AF298810.1 | - | 5 - 6 | 354 | 65 | |
| AK003446.1 | - | 2 - 3 | 70 | 65 | ||
| AK146129.1 | - | 3 - 4 | 204 | 65 | ||
| AK146448.1 | - | 5 - 6 | 436 | 65 | ||
| AK147910.1 | - | 450 | 65 | |||
| AK150981.1 | - | 5 - 6 | 422 | 65 | ||
| AK152367.1 | - | 5 - 6 | 422 | 65 | ||
| AK153250.1 | - | 5 - 6 | 422 | 65 | ||
| AK165761.1 | - | 5 - 6 | 427 | 65 | ||
| BC033457.1 | - | 5 - 6 | 395 | 3991107 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1d
Genomic Map