Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700001557.mm.1 |
TC0700000219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001557.mm.1 |
NM_025898.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001557.mm.1 |
NM_025898.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001558.mm.1 |
TC0700000219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001558.mm.1 |
NM_025898.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001558.mm.1 |
NM_025898.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001562.mm.1 |
TC0700000219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001562.mm.1 |
NM_025898.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001562.mm.1 |
NM_025898.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001559.mm.1 |
TC0700000219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001559.mm.1 |
NM_025898.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001559.mm.1 |
NM_025898.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001560.mm.1 |
TC0700000219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001560.mm.1 |
NM_025898.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001560.mm.1 |
NM_025898.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001561.mm.1 |
TC0700000219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001561.mm.1 |
NM_025898.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001561.mm.1 |
NM_025898.3 |
Refseq |